Learn More
AVANTOR MASTERFLEX (VWR LLC) Tubing, 1/2""ID x 5/8""OD, 25 ft/pack
Catalog No. NC1385020
Encompass
Viton Tubing, 1/2"ID x 5/8"OD, 25 ft/pack, Applications: Acid and solvent transfer; high-temperature work.Characteristics: Opaque black.Certification: Tubing is manufactured according to GMP. Temperature range: ¿25 to 399¿F(¿32 to 204¿C)Sterilize: Not recommended.
Catalog No. NC1385020 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0643407
Viton Tubing, 1/2"ID x 5/8"OD, 25 ft/pack, Applications: Acid and solvent transfer; high-temperature work.Characteristics: Opaque black.Certification: Tubing is manufactured according to GMP. Temperature range: ¿25 to 399¿F(¿32 to 204¿C)Sterilize: Not recommended.