Learn More
AVANTOR MASTERFLEX (VWR LLC) Viton Tubing, 3/16""ID x 1/4""OD, 25 ft/pack
Catalog No. NC9789550
Encompass
Applications: Acid and solvent transfer; high-temperature work. Characteristics: Opaque black. Certification: Tubing is manufactured according to GMP. Temperature range: ¿25 to 399¿F (¿32 to 204¿C) Sterilize: Not recommended.
Catalog No. NC9789550 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0643403
Applications: Acid and solvent transfer; high-temperature work. Characteristics: Opaque black. Certification: Tubing is manufactured according to GMP. Temperature range: ¿25 to 399¿F (¿32 to 204¿C) Sterilize: Not recommended.