Learn More
Julabo USA Inc 1m Viton tubing 10mm ID
SDP
Catalog No. 507534134
Encompass_Preferred
Bath Accessories, Julabo, Viton tubing 1.0m (-35°...+200 °C) 10mm inner diameter. Priced per meter.
Catalog No. 50-753-4134 Supplier Julabo USA Inc Supplier No. 8930110
Bath Accessories, Julabo, Viton tubing 1.0m (-35°...+200 °C) 10mm inner diameter. Priced per meter.