Learn More
Julabo USA Inc 1m Viton tubing 12mm ID
SDP
Catalog No. 507534135
Encompass_Preferred
Bath Accessories, Julabo, Viton tubing 1.0m (-35°...+200 °C) 12mm inner diameter. Priced per meter.
Catalog No. 50-753-4135 Supplier Julabo USA Inc Supplier No. 8930112
Bath Accessories, Julabo, Viton tubing 1.0m (-35°...+200 °C) 12mm inner diameter. Priced per meter.