Learn More
Thermo Scientific VITON TUBING/DRAIN 6/PKVITON
Supplier:  Thermo Scientific 842312051541
Encompass
Pump Drain Tube Organics (Viton); Viton white/white pump tubing suitable for use in the analysis of; organic based samples.; One pack of six tubes.; Compatible with all iCAP 6000 Series Spectrometers.
Catalog No. NC9658742