Learn More
Thermo Scientific VITON TUBING ORGANIC SMPL 6PK
Supplier:  Thermo Scientific 842312051531
Encompass
Pump Sample Tube Organics (Viton); Viton orange/white pump tubing suitable for use in the analysis of; organic based samples.; One pack of six tubes.; Compatible with all iCAP 6000 Series Spectrometers.
Catalog No. NC9658741