Learn More
Perkin Elmer US LLC Viton-PTFE Coated O-Ring, 2.56 mm I.D., for Avio 200/500
Catalog No. 502104176
Encompass_Preferred
This Viton-Teflon coated o-ring can withstand high temperatures and has wide chemical agent compatibility.
Catalog No. 50-210-4176 Supplier Perkin Elmer US LLC Supplier No. 09210635
This Viton-Teflon coated o-ring can withstand high temperatures and has wide chemical agent compatibility.