Learn More
Perkin Elmer US LLC Viton-PTFE Coated O-Ring, 8.74 mm I.D., for Avio 200/500
Catalog No. 502104183
Encompass_Preferred
This Viton-Teflon coated o-ring can withstand high temperatures and has wide chemical agent compatibility.
Catalog No. 50-210-4183 Supplier Perkin Elmer US LLC Supplier No. 09210714
This Viton-Teflon coated o-ring can withstand high temperatures and has wide chemical agent compatibility.