Learn More
Litron Laboratories VITRO MULTIFLOW DNA DAMGE KIT
Catalog No. NC3666106
Encompass
VITRO MULTIFLOW DNA DAMAGE KIT P53, YH2AX, PHOSPHO-HISTONE H3, CLEAVED PARP ALLOWS USERS O ANALYZE IN VITRO SAMPLES FOR DNA DAMAGE USING THEIR OWN FLOW CYTOMETER. THE KIT CONTAINS ALL OF THE QUALITY-ASSURED SUPPLIES AND REAGENTS TO ANALYZE 384 SAMPLES VIA FLOW CYTOMETRY (86 WELL PLATE FORMAT)
Catalog No. NC3666106 Supplier Litron Laboratories Supplier No. MULTFLW P53 YH2AX H3 PARP
VITRO MULTIFLOW DNA DAMAGE KIT P53, YH2AX, PHOSPHO-HISTONE H3, CLEAVED PARP ALLOWS USERS O ANALYZE IN VITRO SAMPLES FOR DNA DAMAGE USING THEIR OWN FLOW CYTOMETER. THE KIT CONTAINS ALL OF THE QUALITY-ASSURED SUPPLIES AND REAGENTS TO ANALYZE 384 SAMPLES VIA FLOW CYTOMETRY (86 WELL PLATE FORMAT)