Learn More
Mitegen Blotting Paper, Vitrobot Compatible, Grade 595, 55mm OD x 20mm ID, For Cryo-EM Sample Preparation, 100 Sheets/Pack
SDP
Catalog No. NC2232529
Encompass
This specialized Blotting Paper is engineered for optimal performance in cryo-electron microscopy (cryo-EM) sample preparation, offering seamless compatibility with the Vitrobot system. Each sheet is crafted from high-quality Grade 595 paper, ensuring consistent and precise blotting for reproducible results. The paper features an outer diameter of 55 mm and an inner diameter of 20 mm, perfectly sized for efficient operation within compatible equipment. Designed to effectively remove excess liquid from samples, this blotting paper is crucial for achieving optimal ice thickness and quality, which are vital for high-resolution cryo-EM imaging. Its reliable performance contributes to minimizing sample waste and accelerating your experimental workflow. This essential consumable is supplied in a convenient pack of 100 sheets, providing ample supply for demanding laboratory needs in cryo-EM.
This item is not returnable. View return policy
Catalog No. NC2232529 Supplier Mitegen Supplier No. MCEMVBBP100
This specialized Blotting Paper is engineered for optimal performance in cryo-electron microscopy (cryo-EM) sample preparation, offering seamless compatibility with the Vitrobot system. Each sheet is crafted from high-quality Grade 595 paper, ensuring consistent and precise blotting for reproducible results. The paper features an outer diameter of 55 mm and an inner diameter of 20 mm, perfectly sized for efficient operation within compatible equipment. Designed to effectively remove excess liquid from samples, this blotting paper is crucial for achieving optimal ice thickness and quality, which are vital for high-resolution cryo-EM imaging. Its reliable performance contributes to minimizing sample waste and accelerating your experimental workflow. This essential consumable is supplied in a convenient pack of 100 sheets, providing ample supply for demanding laboratory needs in cryo-EM.