Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ VitroEase™ Apoferritin Standard

Catalog No. PIA51362 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 x 20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA51362 5 x 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIA51362 Supplier Thermo Scientific™ Supplier No. A51362
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Thermo Scientific™ VitroEase™ Apoferritin Standard is a highly validated pure protein for use as a quality control sample for cryo-electron microscopy single particle analysis.

  • A highly validated pure protein for use as a quality control sample for cryo-electron microscopy single particle analysis
  • Designed specifically to meet the stringent needs for high protein purity, minimal protein aggregation, and intact protein structure
  • Protein sample grid preparation is a major bottleneck in cryo-electron microscopy (cryo-EM) single particle analysis (SPA), often requiring days or even weeks to optimize
  • Apoferritin, a 24-subunit protein (474 kDa), is considered the gold standard for cryo-EM SPA
  • Protein production is optimized and validated for use in cryo-EM grid preparation and analysis
  • High-quality, consistent protein standard documented via lot-specific certificates of analysis
  • Ready-to-use format with each single-use vial sufficient for 5 to 6 grid preparations
  • Standard is fit for use as a comparison for success for new users, or as quality control for expert users
TRUSTED_SUSTAINABILITY

Specifications

Concentration 3.5–4.0 mg/mL
For Use With (Application) Cryo-EM Single Particle Analysis (SPA)
Name Apoferrtin
Quantity 5 x 20 μL
Gene Alias ApoF
Gene Symbol FTH1, FTH
Sterility Non-sterile
Detection Method Cryo-EM
For Use With (Equipment) Cryo-Electron Microscopes, Mass spectrometers
Sample Type Purified protein
Technique MS, Cryo-EM SPA
Volume (Metric) 100 μL
Expression System Bacterial
Form Liquid
Color Colorless
Ligand Type Iron
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.