Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ VitroEase™ Buffer Screening Kit and Detergents

Catalog No. FERA65377 Shop All Thermo Scientific Products
Change view
Click to view available options
Buffer:
CHAPS
FOM
OG
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Buffer
FERA65377 FOM
FERA65373 CHAPS
FERA65374 OG
FERA49856 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. FERA65377 Supplier Thermo Scientific™ Supplier No. A65377
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The Thermo Scientific VitroEase Buffer Screening Kit provides pre-formulated buffers and detergents and a comprehensive screening strategy to identify optimal protein conditions for single particle cryo-EM sample analysis.

The Thermo Scientific VitroEase Buffer Screening Kit provides pre-formulated buffers and detergents and a comprehensive screening strategy to identify optimal protein conditions for single particle cryo-EM sample analysis.

Features of the VitroEase Buffer Screening Kit include:
• Ease of use—14 buffers and 6 detergents pre-formulated at 10X concentration ready to be spiked in with your protein sample with minimal sample dilution
• Convenient—buffers and detergents provided with color-coded caps that correlate with color-coded screening strategy
• Comprehensive—optimized screening strategy encompassing two rounds of screening with 24 grids with trouble-shooting suggestions
• Flexible—wide range of common buffers and detergents for cryo-EM sample preparation sufficient for sample optimization of 400 grids
• Validated—components and screening strategy validated with single particle cryo-EM analysis

Cryo-EM sample screening evaluates protein behavior in various buffers and additive conditions with the end goal of identifying the optimal conditions in which the protein is stable, randomly oriented, and equally distributed in ice. Kit buffers are provided with pHs from 3.6 to 8.9 pre-mixed with various salts and divalent cations. Detergents are provided at 10X concentrations with an easy-to-use table to convert volume to CMC concentration.

The comprehensive sample screening strategy uses two rounds of screening and 24 grids, taking approximately two days to process (figure below). In the first round, optimal protein concentration, buffer, and pH conditions are determined. In the second round, detergents are further optimized. A condensed strategy is also provided using 8 grids and taking one day to process, which can be used when little sample is available or for use with side entry or semi-automated sample loading systems, like the Tundra Cryo-TEM.

The recommended screening strategies are designed for use with the Thermo Scientific Vitrobot Mk4 and Krios, Glacios, Arctica microscopes with Autoloader systems and EPU multigrid.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Detergent F (refill)
Physical Form Liquid
Concentration 0.7% (10X CMC)
Quantity 2 x 100 μL
Packaging Polypropylene Vials
Buffer FOM
For Use With (Equipment) Vitrobot™ Mark IV System, Krios™, Glacios™,or Talos™ Arctica™ Microscopes

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.