Learn More
Enzo Life Sciences Vitronectin (human), (recombinant) (500µg)
SDP
Catalog No. 502013966
Encompass_Preferred
Alternative name: VTN, Serum-spreading factor, V75. Purity: ≥95% (SDS & HPLC). Formulation: Lyophilized from 20mM sodium phosphate, pH 7.5. Source: Produced in HEK 293 cells. Long Term Storage: -20°C.
Catalog No. 50-201-3966 Supplier Enzo Life Sciences Supplier No. ENZPRT1790500
Alternative name: VTN, Serum-spreading factor, V75. Purity: ≥95% (SDS & HPLC). Formulation: Lyophilized from 20mM sodium phosphate, pH 7.5. Source: Produced in HEK 293 cells. Long Term Storage: -20°C.