Learn More
Advanced Biomatrix Vitronectin, Human, 0.1 mg
Catalog No. 501960274
Encompass_Preferred
Vitronectin is a monomeric glycoprotein used to promote cell attachment, migration, proliferation and differentiation in a broad number of cell lines and types. This product has been purified from human plasma where it is found as a mixture of 75kDa and 65kDa polypeptides.Vitronectins primary use in cell culture is related to cell adhesion. It also binds to heparin and collagen.Vitronectin is ideal for coating of surfaces. The optimal concentration for cell attachment and culture may differ for various cell types. Vitronectin has been used at a final coating concentration as low as 50 ng/cm2 on plasticware. It is provided in user-friendly packaging for use and storage. Vitronectin is sterile filtered and is supplied as a ready to use solution after thawing and concentration adjustment. This product is shipped separately on dry ice.
Catalog No. 50-196-0274 Supplier Advanced Biomatrix Supplier No. 50510.1MG
Vitronectin is a monomeric glycoprotein used to promote cell attachment, migration, proliferation and differentiation in a broad number of cell lines and types. This product has been purified from human plasma where it is found as a mixture of 75kDa and 65kDa polypeptides.Vitronectins primary use in cell culture is related to cell adhesion. It also binds to heparin and collagen.Vitronectin is ideal for coating of surfaces. The optimal concentration for cell attachment and culture may differ for various cell types. Vitronectin has been used at a final coating concentration as low as 50 ng/cm2 on plasticware. It is provided in user-friendly packaging for use and storage. Vitronectin is sterile filtered and is supplied as a ready to use solution after thawing and concentration adjustment. This product is shipped separately on dry ice.