Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Vitronectin (VTN-N) Recombinant Human Protein, truncated

Catalog No. A400457
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
1 mL
10 mL
Packaging:
Tubes
Vial
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Packaging
A400457 1 mL Tubes
A400458 10 mL Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A400457 Supplier Gibco™ Supplier No. A400457
Get 15% Off Cell Culture Products
Use promo code CELLS15
View all promotions
Add to Cart
Edge
Add to Cart

Vitronectin (VTN-N) is a recombinant human protein that provides a defined surface for feeder-free culture of human pluripotent stem cells (PSCs).

Vitronectin (VTN-N) is a recombinant human protein that offers a defined surface for feeder-free culture of human pluripotent stem cells (PSCs). When used with Essential 8 Medium (Cat. No. A1517001), vitronectin has been proven to maintain pluripotency and normal growth characteristics in multiple PSC lines. Vitronectin (VTN-N) has been shown to support PSC growth for >50 passages without any signs of karyotypic abnormalities and to maintain the ability of PSCs to differentiate into all three germ line lineages.

Vitronectin (VTN-N) is:

  • Optimized—designed in the laboratory of James Thomson for use with Essential 8 Medium
  • Defined—for reduced variability in your PSC cultures
  • Cost effective—for economical and scalable PSC culture

Optimized and defined surface

Unlike standard basement membrane extracts (BMEs), vitronectin (VTN-N) is based on the N-variant design of vitronectin from the laboratory of James Thomson for use with Essential 8 Medium. The vitronectin (VTN-N) variant has been shown to support human pluripotent stem cell attachment and survival better than wild type vitronectin. Vitronectin (VTN-N) helps provide an optimal environment for feeder-free pluripotent stem cell culture.

Reduced variability

Vitronectin (VTN-N) is a defined, recombinant human protein that reduces variability in your PSC cultures compared to human-plasma derived vitronectin and standard basement membrane extracts (BMEs).

Cost effective

When used with Essential 8 Medium, vitronectin (VTN-N) provides a cost effective, defined system for feeder free culture of human pluripotent stem cells (PSCs).

Commercialized in Partnership with Cellular Dynamics International.

Reference

(1) Chen G, et al. Chemically defined conditions for human iPSC derivation and culture. Nat Methods. 2011 8(5):424-9.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.81 to 0.99 mg/mL
For Use With (Application) Pluripotent Stem Cell Culture
Quantity 1 mL = 0.9 mg
Packaging Tubes
Product Type Pluripotent Stem Cell Matrix
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Sterility Sterile-filtered
Content And Storage Store at -70°C
Classification Animal Origin-Free
Shipping Condition Dry Ice
Volume (Metric) 1 mL
Form Liquid
Color Clear
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.