Learn More
STEMCELL Technologies Vitronectin XF™, Size: 2 mL
SDP
Catalog No. NC0672882
Encompass
Defined, xeno-free matrix that supports the growth and differentiation of human pluripotent stem cells under serum-free, feeder-free conditions. 1 x 2 mL vial.
Catalog No. NC0672882 Supplier STEMCELL Technologies Supplier No. 07180
Defined, xeno-free matrix that supports the growth and differentiation of human pluripotent stem cells under serum-free, feeder-free conditions. 1 x 2 mL vial.