Learn More
THEWELL BIOSCIENCE VITROPRIME 3D CULTURE PLT 24W
Catalog No. NC3974151
Encompass
A premium cover-glass bottom plate for zero disruption 3D cell culture workflow: From cell seeding to high-resolution imaging. 24 well.
Catalog No. NC3974151 Supplier THEWELL BIOSCIENCE Supplier No. VP3D24W5
A premium cover-glass bottom plate for zero disruption 3D cell culture workflow: From cell seeding to high-resolution imaging. 24 well.