Learn More
THEWELL BIOSCIENCE THEWELL BIOSCIENCE 3D Culture and Imaging Plate, 6-Well, Cover-Glass, Anti-Floating
Supplier:  THEWELL BIOSCIENCE VP3D6W5
Encompass
The VitroPrime™ 3D Culture and Imaging Plate is a premium 3D cell culture plate with unique media-exchange channels, “sample-locking” technology, and an ultra-clear cover-glass bottom for zero-disruption 3D cell culture workflow.
  • Anti-floating and anti-rotation technology prevents sample loss and “drifting” during medium changes and imaging.
  • Disturbance-free medium exchange via dedicated channels ensures smooth nutrient diffusion without disrupting 3D hydrogel.
  • End-to-end workflow integration supports complete “in-plate” processes from embedding to imaging, minimizing sample loss.
  • Elite optical clarity with a premium #1.5 cover-glass bottom offers maximum light transmission and minimal auto-fluorescence for high-resolution imaging.
  • Automation ready and compatible with automated liquid-handling systems and high-content imaging platforms.
Catalog No. NC3974156