Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho Vitros 3600 Immunodiagnostic System

Catalog No. OCD6802914
Change view
Click to view available options
Depth (English):
33.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Depth (English)
OCD6802914 33.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OCD6802914 Supplier QuidelOrtho Supplier No. 6802914
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Delivers high-quality results, with a comprehensive testing menu covering the major disease states.

  • Meet your lab's immunoassay testing needs with the VITROS 3600 system, designed to deliver results with speed and accuracy
  • Equipped with built-in intelligent technologies, the VITROS 3600 has the quality and reliability that comes from constant innovation and improvement
  • Rely on accurate results: 85% of VITROS™ assays are designated World Class or Excellent as defined by Six Sigma - the most in the industry
  • Simplify with a waterless lab: Because VITROS systems do not rely on water, you have greater flexibility to plan your lab layout
  • Save time: Expect greater time savings with a first-pass yield of 96.5%, complete the analytical process the first time without intervention or repeat testing
  • Maximize uptime: VITROS systems are ready when you are with industry-leading analyzer uptime, minimal daily maintenance and 24-hour ready mode

Specifications

Type Automated Enzyme Immunoassay Analyzer
Depth (English) 33.5 in.
Depth (Metric) 0.85 m
Height (English) 64.5 in.
Height (Metric) 1.64 m
Width (English) 83.5 in.
Width (Metric) 2.12 m
Weight (English) 1580 lb.
Weight (Metric) 717 kg
Dimensions (L x W x H) 33.5 x 83.5 x 64.5 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.