Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
- Home
- Products
- Research and Clinical Diagnostic Tests and Controls
- Chemistry Controls, Calibrators and Reagents
- Clinical Chemistry Reagents
- QuidelOrtho VITROS™ MicroTip™ Supplies Performance Verifiers
QuidelOrtho VITROS™ MicroTip™ Supplies Performance Verifiers
Catalog No. OCD6801751
Change view
Click to view available options
Quantity:
3 Pack
6 Pack
Type:
Performance Verifier I
Performance Verifier II
Performance Verifier III
For Use With (Application):
Antistreptolysin, Rheumatoid Factor
ApoA1
ApoB
Direct Total Iron Binding Capacity
High Sensitivity C-Reactive Protein
Microalbumin
Prealbumin
Protein
alpha-1-antitrypsin, Haptoglobin Reagent
19 product options available for selection
| Catalog No. | Quantity | Type | For Use With (Application) |
|---|---|---|---|
| OCD6801751 | 3 Pack | Performance Verifier II | Protein |
| OCD6801771 | 6 Pack | Performance Verifier I | Prealbumin |
| OCD6801772 | 6 Pack | Performance Verifier II | Prealbumin |
| OCD6801769 | 6 Pack | Performance Verifier I | ApoA1 |
| OCD6801744 | 6 Pack | Performance Verifier I | Protein |
| OCD6801745 | 6 Pack | Performance Verifier II | Protein |
| OCD6802312 | 6 Pack | Performance Verifier II | alpha-1-antitrypsin, Haptoglobin Reagent |
| OCD6801750 | 3 Pack | Performance Verifier I | Protein |
| OCD6802411 | 6 Pack | Performance Verifier I | Antistreptolysin, Rheumatoid Factor |
| OCD6802311 | 6 Pack | Performance Verifier I | alpha-1-antitrypsin, Haptoglobin Reagent |
| OCD6802412 | 6 Pack | Performance Verifier II | Antistreptolysin, Rheumatoid Factor |
| OCD6801742 | 6 Pack | Performance Verifier I | High Sensitivity C-Reactive Protein |
| OCD6801747 | 6 Pack | Performance Verifier II | Microalbumin |
| OCD6801770 | 6 Pack | Performance Verifier I | ApoB |
| OCD6802313 | 6 Pack | Performance Verifier III | alpha-1-antitrypsin, Haptoglobin Reagent |
| OCD6801768 | 6 Pack | Performance Verifier III | Protein |
| OCD6801746 | 6 Pack | Performance Verifier I | Microalbumin |
| OCD6802349 | 6 Pack | Performance Verifier II | Direct Total Iron Binding Capacity |
| OCD6802348 | 6 Pack | Performance Verifier I | Direct Total Iron Binding Capacity |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
Specifications
Specifications
| Type | Performance Verifier II |
| For Use With (Application) | Protein |
| Quantity | 3 Pack |
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?