Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho Vitros™ XT 7600 Integrated System

Catalog No. 23610201
Change view
Click to view available options
Sample Size:
2 to 80 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Size
23-610-201 2 to 80 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-610-201 Supplier QuidelOrtho Supplier No. 6844461

This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Vitros XT 7600 Integrated system offers a full clinical chemistry and immunoassay menu in one easy-to-use system.

The Vitros XT 7600 Integrated system offers a full clinical chemistry and immunoassay menu in one easy-to-use system that delivers industry-leading uptime (Data on file). Built-in technologies improve efficiency and quality, allowing for scalable automation solutions that streamline your workflow.

When faced with higher volumes and fewer resources, your lab needs a smarter way to work. Our flagship Vitros™ offers the highest level of reliability and quality to make it easier to focus on doing more with less.

Combining innovation and scale, this truly integrated system can process a test every three seconds with exceptional accuracy. The system accommodates more than 160 tests to meet all your needs today, with the capacity to expand your menu to meet the demands of tomorrow. It's time to introduce your lab to the future and discover what's possible.

Specifications

Depth (English) 34.9 in.
Operating Temperature Range (English) 15°C to 30°C
Operating Temperature Range (Metric) 59°F to 86°F
Product Type Immunodiagnostic System
Frequency 47/63 Hz
Depth (Metric) 0.89 m
Height (English) 68 in.
Height (Metric) 1.73 m
Sample Size 2 to 80 μL
Sample Type Serum, Plasma, Urine, CSF, Whole Blood (% Hemoglobin A1c and Red Cell Folate)
Voltage 200/240 V
Weight (English) 2360 lb.
Weight (Metric) 1070.5 kg
Width (English) 110 in.
Width (Metric) 2.79 m
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.