Learn More
VITRO VIVO BIOTECH LLC VitroView™ Luxol Fast Blue Stain Kit
Catalog No. NC3832496
Encompass_Preferred
Luxol Fast Blue StainKit is designed for staining myelin/myelinated axons on formalin-fixed, paraffin-embedded brain and spinal cord tissue sections, as well as frozen sections. The myelin, including phospholipids, will be stained blue to green, and the neurons will be stained violet. This stain is commonly used for identifying the basic neuronal structure in brain or spinal cord tissue.(A printed user manual is included)VB-3006-1 Luxol Fast Blue Solution 250 mlVB-3006-2 Cresyl Violet Solution 250 mlVB-3006-3 Lithium Carbonate Solution 250 ml
Catalog No. NC3832496 Supplier VITRO VIVO BIOTECH LLC Supplier No. VB3006
Luxol Fast Blue StainKit is designed for staining myelin/myelinated axons on formalin-fixed, paraffin-embedded brain and spinal cord tissue sections, as well as frozen sections. The myelin, including phospholipids, will be stained blue to green, and the neurons will be stained violet. This stain is commonly used for identifying the basic neuronal structure in brain or spinal cord tissue.(A printed user manual is included)VB-3006-1 Luxol Fast Blue Solution 250 mlVB-3006-2 Cresyl Violet Solution 250 mlVB-3006-3 Lithium Carbonate Solution 250 ml