Learn More
VITRO VIVO BIOTECH LLC VitroView™ Nissl Stain Kit
Catalog No. NC3832500
Encompass_Preferred
A Nissl body, also known as Nissl or tigroid substance, is a large granular body found in neurons. This Nissl Stain Kit is designed for the detection of Nissl body in the cytoplasm of neurons on paraformaldehyde fixed frozen or vibratome tissue sections. This stain is commonly used for identifying the basic neuronal structure in brain or spinal cord tissue. The Nissl body will be stained purple-blue.(A printed user manual is included)VB-3010-1 Cresyl Violet Solution 250 ml VB-3010-2 Alcohol/chloroform Solution 250ml
Catalog No. NC3832500 Supplier VITRO VIVO BIOTECH LLC Supplier No. VB3010
A Nissl body, also known as Nissl or tigroid substance, is a large granular body found in neurons. This Nissl Stain Kit is designed for the detection of Nissl body in the cytoplasm of neurons on paraformaldehyde fixed frozen or vibratome tissue sections. This stain is commonly used for identifying the basic neuronal structure in brain or spinal cord tissue. The Nissl body will be stained purple-blue.(A printed user manual is included)VB-3010-1 Cresyl Violet Solution 250 ml VB-3010-2 Alcohol/chloroform Solution 250ml