Learn More
VITRO VIVO BIOTECH LLC VitroView™ PAS Stain Kit
Catalog No. NC3832495
Encompass_Preferred
The Periodic Acid Schiff or PAS Stain Kit is used for detection of glycogen in tissues such as liver, cardiac and skeletal muscle in formalin-fixed, paraffin-embedded tissue sections. The PAS stain kit may also be used for frozen sections. The PAS reaction in tissue sections is useful for the demonstration of mucopolysaccharides. PAS staining may also be used for the demonstration of fungal organisms in tissue sections. Glycogen, mucin, and fungi will stain purple and nuclei will stain blue. (A printed user manual is included)VB-3004-1 Periodic Acid Solution 250 mlVB-3004-2 Schiff reagent 250 ml VB-3004-3 Mayer’s Hematoxylin Solution 250ml
Catalog No. NC3832495 Supplier VITRO VIVO BIOTECH LLC Supplier No. VB3004
The Periodic Acid Schiff or PAS Stain Kit is used for detection of glycogen in tissues such as liver, cardiac and skeletal muscle in formalin-fixed, paraffin-embedded tissue sections. The PAS stain kit may also be used for frozen sections. The PAS reaction in tissue sections is useful for the demonstration of mucopolysaccharides. PAS staining may also be used for the demonstration of fungal organisms in tissue sections. Glycogen, mucin, and fungi will stain purple and nuclei will stain blue. (A printed user manual is included)VB-3004-1 Periodic Acid Solution 250 mlVB-3004-2 Schiff reagent 250 ml VB-3004-3 Mayer’s Hematoxylin Solution 250ml