Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ VITLAB™ VITsafe™ Wide-Mouth Wash Bottle

Catalog No. 14380961 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Chemical Label:
Acetone
Distilled Water
Ethanol
Heptane
Xylene
i-Hexane
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Label
14-380-961 Acetone
14-380-960 Ethanol
14-380-956 Heptane
14-380-957 i-Hexane
14-380-959 Distilled Water
14-380-958 Xylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-380-961 Supplier BRANDTECH™ VITLAB™ Supplier No. 1452829
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Wide-Mouth Wash Bottle with integrated vent in cap to reduce dripping.

  • Chemical name in German, English, French and Spanish
  • Chemical formula
  • CAS Number, hazard pictogram, signal word
  • Risk Phrases (H phrases), safety phrases (P phrases)
  • NFPA Code
  • Fine tip polypropylene tube provides directional accuracy
  • Colored cap allows easier identification
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 500 mL
Color Red
For Use With (Application) Acetone
Material Polypropylene
Length (Metric) 166 mm
Packaging Pack
Quantity 12
Sterility Non-sterile
Width (Metric) 58 mm
Closure Included Yes
Graduated No
Mouth Wide, Threaded
Height (Metric) 166 mm
Length (English) 6.54 in.
Height (English) 6.54 in.
Chemical Label Acetone
Closure Size GL 45
Diameter (Metric) 76 mm
Closure Type Spouted Cap
Through the Closure Dispensing Vented Tube
Autoclavable Not Autoclavable
Capacity (English) 16.9 oz.
Shape Cylindrical
Disposable Reusable
Product Type Safety-labeled Wash Bottle
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.