Learn More
MSC VITURA SAFETY GLASSES
Catalog No. 5000789202
Encompass
These safety glasses are designed for high-visibility applications, offering protection against impacts and harmful UV radiation. They feature a durable polycarbonate construction and advanced lens coatings to maintain clear vision in challenging environments.
  • Anti-fog coatings improve visibility under hot and humid conditions
  • Removable, foam-lined gasket helps limit eye exposure to nuisance dust while providing added comfort and cushioning
  • Small vents in the gasket help minimize fogging
  • Corded earplug control system designed to keep eyewear and earplugs attached, untangled and ready to use
  • Polycarbonate lenses absorb 99.9% UVA and UVB
Catalog No. 50-007-89202 Supplier MSC Supplier No. 70452883
These safety glasses are designed for high-visibility applications, offering protection against impacts and harmful UV radiation. They feature a durable polycarbonate construction and advanced lens coatings to maintain clear vision in challenging environments.
  • Anti-fog coatings improve visibility under hot and humid conditions
  • Removable, foam-lined gasket helps limit eye exposure to nuisance dust while providing added comfort and cushioning
  • Small vents in the gasket help minimize fogging
  • Corded earplug control system designed to keep eyewear and earplugs attached, untangled and ready to use
  • Polycarbonate lenses absorb 99.9% UVA and UVB