Learn More
Viva Products Vivacon 2-PCR Grade Centrifugal Concentrator (ETO treated)
SDP
Supplier:  Viva Products VN02H42ETO
Encompass_Preferred
Centrifugal Ultrafiltration Device; Vivacon Model 2; For DNA concentration and desalting; Patented Hydrosart regenerated cellulose membrane for high recovery; Treated with dual-cycle ETO gas process to sterilize and denature DNA; Sample volume to 2 ml; Final concentrate volume as low as 55 microliters; Reverse spin for easy concentrate recovery; Fits fixed angle rotors for 15 ml (17 mm) conical bottom tubes; 125 mm length; 100,000 MWCO; 100/PK
Catalog No. 50-311-835