Learn More
Viva Products Vivacon 500-PCR Grade Centrifugal Concentrator (ETO treated)
SDP
Supplier:  Viva Products VN01H22ETO
Encompass_Preferred
Centrifugal Ultrafiltration Device; Vivacon Model 500; For DNA concentration and desalting; Patented Hydrosart regenerated cellulose membrane for high recovery; Treated with dual-cycle ETO gas process to sterilize and denature DNA; Sample volume to 0.5 ml; Final concentrate volume as low as 5 microliters; Reverse spin for easy concentrate recovery; Fits fixed angle rotors for 1.5/2.2 ml (11 mm) conical bottom tubes; 47.5 mm length; 30,000 MWCO; 100/PK
Catalog No. 50-311-816