Learn More
Viva Products Vivaspin 4 Centrifugal Concentrator
SDP
Catalog No. 50355731
Encompass_Preferred
Centrifugal Ultrafiltration Concentrator; Vivaspin Model 4; For concentration and desalting of proteins and biologicals; Useful for clinical samples such as urine and spinal fluid; Used prior to electrophoresis of monoclonal immunoglobulins (M-proteins) as well as immunofixation electrophoresis (IFE); Sample volume to 4 ml; Final concentrate volume as low as 20 microliters for 200X concentration factor; Fits fixed angle and swinging bucket rotors for 15 ml (17 mm diameter) conical bottom tubes; 122 mm length; 10,000 MWCO; 100/PK
Catalog No. 50-355-731 Supplier Viva Products Supplier No. VS0404
Centrifugal Ultrafiltration Concentrator; Vivaspin Model 4; For concentration and desalting of proteins and biologicals; Useful for clinical samples such as urine and spinal fluid; Used prior to electrophoresis of monoclonal immunoglobulins (M-proteins) as well as immunofixation electrophoresis (IFE); Sample volume to 4 ml; Final concentrate volume as low as 20 microliters for 200X concentration factor; Fits fixed angle and swinging bucket rotors for 15 ml (17 mm diameter) conical bottom tubes; 122 mm length; 10,000 MWCO; 100/PK