Learn More
Viva Products Vivaspin 4 Centrifugal Concentrator
SDP
Supplier:  Viva Products VS0403
Encompass_Preferred
Centrifugal Ultrafiltration Concentrator; Vivaspin Model 4; For concentration and desalting of proteins and biologicals; Useful for clinical samples such as urine and spinal fluid; Used prior to electrophoresis of monoclonal immunoglobulins (M-proteins) as well as immunofixation electrophoresis (IFE); Sample volume to 4 ml; Final concentrate volume as low as 20 microliters for 200X concentration factor; Fits fixed angle and swinging bucket rotors for 15 ml (17 mm diameter) conical bottom tubes; 122 mm length; 10,000 MWCO; 25/PK
Catalog No. 50-355-730