Learn More
Biotium VivoBrite™ Rapid Antibody Labeling Kit for Small Animal In Vivo Imaging, CF770 SE
Catalog No. 501964909
Encompass_Preferred
This labeling kit comprises our superior near-IR amine-reactive CF dye and other components necessary for carrying out antibody labeling, purification, and filter sterilization. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Near-infrared fluorescent CF770 dye has excitation/emission at 770/797 nm.
Catalog No. 50-196-4909 Supplier Biotium Supplier No. 92162
This labeling kit comprises our superior near-IR amine-reactive CF dye and other components necessary for carrying out antibody labeling, purification, and filter sterilization. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Near-infrared fluorescent CF770 dye has excitation/emission at 770/797 nm.