Learn More
American Research Products Inc VivoGenie AntiHuman CD4
Catalog No. 5000304844
Encompass
VivoGenie Anti-Human CD4 (Ibalizumab) Biosimilar is a vivogenie with human reactivity Available in 1mg size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles
Catalog No. 50-003-04844 Supplier American Research Products Inc Supplier No. IVMB04991MG
VivoGenie Anti-Human CD4 (Ibalizumab) Biosimilar is a vivogenie with human reactivity Available in 1mg size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles