Learn More
American Research Products Inc VivoGenie AntiHuman CD47
Catalog No. 5000306974
Encompass
VivoGenie Anti-Human CD47 (Magrolimab) Biosimilar is a vivogenie with human reactivity Available in 1mg size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles
Catalog No. 50-003-06974 Supplier American Research Products Inc Supplier No. IVMB040350MG
Add to Cart
Add to Cart
Request a Quote
VivoGenie Anti-Human CD47 (Magrolimab) Biosimilar is a vivogenie with human reactivity Available in 1mg size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles