Learn More
American Research Products Inc VivoGenie AntiHuman Nectin4
Supplier:  American Research Products Inc IVMB0409100MG
Encompass
VivoGenie Anti-Human Nectin-4 (Enfortumab) Biosimilar is a vivogenie with human reactivity Available in 100mg size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles
Catalog No. 50-003-06177
Add to Cart
Request a Quote