Learn More
American Research Products Inc VivoGenie AntiHuman PDL1
Catalog No. 5000306570
Encompass
VivoGenie Anti-Human PD-L1 (CD274) (Durvalumab) is a vivogenie with human reactivity produced in human Available in 1 mg size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles
Catalog No. 50-003-06570 Supplier American Research Products Inc Supplier No. IVMB040250MG
Add to Cart
Add to Cart
Request a Quote
VivoGenie Anti-Human PD-L1 (CD274) (Durvalumab) is a vivogenie with human reactivity produced in human Available in 1 mg size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles