Learn More
American Research Products Inc VivoGenie AntiTNF alpha
Supplier: American Research Products Inc IVMB02031MG
VivoGenie Anti-TNF alpha (Adalimumab) Biosimilar is a vivogenie with human reactivity Available in 100 mg size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles