Learn More
American Research Products Inc VivoGenie SARSCoV2 Spike RBD
Catalog No. 5000304363
Encompass
VivoGenie SARS-CoV-2 Spike RBD [2381] Low Endotoxin Antibody is a vivogenie with sars-cov-2 virus reactivity Available in 100 ug size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles
Catalog No. 50-003-04363 Supplier American Research Products Inc Supplier No. IVMB0233100UG
VivoGenie SARS-CoV-2 Spike RBD [2381] Low Endotoxin Antibody is a vivogenie with sars-cov-2 virus reactivity Available in 100 ug size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles