Learn More
SOMNI Scientific™ VivoTherm Induction Chamber Heater
SDP
Catalog No. 04731228 Shop All Somni Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Large Induction Chamber
Medium Induction Chamber
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
04-731-228 Medium Induction Chamber
04-731-226 Large Induction Chamber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 04-731-228 Supplier SOMNI Scientific™ Supplier No. HT15007
Add to Cart
Add to Cart
Deliver consistent, uniform heat to help counteract rapid heat loss commonly observed during isoflurane induction
  • Designed to integrate seamlessly with standard induction chambers (150 x 150 x 300 mm or smaller)
  • Minimizes hypothermia-related variability
  • Supports more reliable physiology and improved experimental consistency
  • Maintain animal body temperature
  • Improve anesthetic stability
  • Built for routine use in research environments
  • Compatible with standard cleaning protocols
  • A purpose-built solution for controlled warming during induction
Specifications
Autoclavable Not Autoclavable
Animal Mouse, Rat
Type Chamber Heater
Disposable Reusable
For Use With (Equipment) Medium Induction Chamber
Size Medium
Sterility Non-sterile
Voltage 120 V