Learn More
Medchemexpress LLC VK-II-86 | 955371-84-3 | 93.3% | 420.50 | 500 MG
SDP
Catalog No. 5000343666
Encompass_Preferred
VK-II-86 is a Carvedilol analogue that lacks antagonist activity at β-adrenoceptors in hypokalaemia. This compound effectively prevents hypokalaemia-induced ventricular arrhythmia through its multi-channel effects, counteracting all hypokalaemia-induced changes in ion channel activity and oxidative stress. For research use only.
  • Carvedilol analogue
  • Lacks antagonist activity at β-adrenoceptors
  • Prevents hypokalaemia-induced ventricular arrhythmia
  • Exhibits multi-channel effects
  • Counters hypokalaemia-induced changes in ion channel activity
  • Counters hypokalaemia-induced oxidative stress
  • Markedly reduces non-sustained and sustained ventricular tachyarrhythmias in a hypokalaemic murine whole-heart model
Catalog No. 50-003-43666 Supplier Medchemexpress LLC Supplier No. HY1110131MG
VK-II-86 is a Carvedilol analogue that lacks antagonist activity at β-adrenoceptors in hypokalaemia. This compound effectively prevents hypokalaemia-induced ventricular arrhythmia through its multi-channel effects, counteracting all hypokalaemia-induced changes in ion channel activity and oxidative stress. For research use only.
  • Carvedilol analogue
  • Lacks antagonist activity at β-adrenoceptors
  • Prevents hypokalaemia-induced ventricular arrhythmia
  • Exhibits multi-channel effects
  • Counters hypokalaemia-induced changes in ion channel activity
  • Counters hypokalaemia-induced oxidative stress
  • Markedly reduces non-sustained and sustained ventricular tachyarrhythmias in a hypokalaemic murine whole-heart model