Learn More
Medchemexpress LLC VK3-OCH3 | 255906-59-3 | 98.64% | 262.33 | 10 MG
SDP
Catalog No. 5000452604
Encompass_Preferred
VK3-OCH3 is a potent antitumor agent that demonstrates cytotoxicity for neuroblastoma cell lines while exhibiting low cytotoxicity for normal cell lines. It induces apoptosis and cell cycle arrest at the G2/M phase in IMR-32 cells and shows antitumor activity. This product is for research use only and not for sale to patients.
  • Potent antitumor activity
  • Selectively cytotoxic to neuroblastoma cell lines
  • Low cytotoxicity for normal cell lines
  • Induces apoptosis
  • Causes cell cycle arrest at G2/M phase in IMR-32 cells
Catalog No. 50-004-52604 Supplier Medchemexpress LLC Supplier No. HYW01176210MG
VK3-OCH3 is a potent antitumor agent that demonstrates cytotoxicity for neuroblastoma cell lines while exhibiting low cytotoxicity for normal cell lines. It induces apoptosis and cell cycle arrest at the G2/M phase in IMR-32 cells and shows antitumor activity. This product is for research use only and not for sale to patients.
  • Potent antitumor activity
  • Selectively cytotoxic to neuroblastoma cell lines
  • Low cytotoxicity for normal cell lines
  • Induces apoptosis
  • Causes cell cycle arrest at G2/M phase in IMR-32 cells