Learn More
TARGETMOL CHEMICALS INC VK3-OCH3 5MG
SDP
Catalog No. 502877808
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.VK3-OCH3 (2-[(2-Methoxy)ethylthio]-3-methyl-1,4-na) is a selective antitumor agent via heme oxygenase (HO-1) related mechanisms; Vitamine K3 analogue. Purity 98%
Catalog No. 50-287-7808 Supplier TARGETMOL CHEMICALS INC Supplier No. T34595MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.VK3-OCH3 (2-[(2-Methoxy)ethylthio]-3-methyl-1,4-na) is a selective antitumor agent via heme oxygenase (HO-1) related mechanisms; Vitamine K3 analogue. Purity 98%