Learn More
MCR Safety™ VL2 MAXBLUE Technology Lenses Safety Glasses
Catalog No. 19827920 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Lens Coating:
Scratch-resistant, Impact-resistant
Catalog No. Lens Coating
19-827-920 Scratch-resistant, Impact-resistant
1 options
Catalog No. 19-827-920 Supplier MCR Safety™ Supplier No. VL210MB
VL2 Series blue light safety glasses - MAXBLUE™ lens technology, navy blue frame, soft secure TPR nose piece.
  • Allows 88% of visible light
  • Athletic inspired design
  • Comfortable TPR nosepieces
  • Hybrid temple design
  • Lens technology blocks 41% of ISO 12312-1 blue light
  • Streamlined 3-point ratcheting temple
  • Meets ANSI/ISEA Z87.1 2020 standard
Specifications
Lens Coating Scratch-resistant, Impact-resistant
Lens Tint Clear
Product Type Glasses
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection U6
Frame Type Half Frame
Autoclavable Not Autoclavable