Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC VL285 Phenol | 1448188-69-9 | C29H32N4O5S | 25 MG
SDP

Catalog No. 5000374421
Encompass_Preferred

VL285 Phenol is a phenol-based VL285 analog and an E3 ubiquitin ligase VHL ligand. It can be used in the synthesis of HaloPROTAC3, where it aids in degrading HaloTag7 fusion protein with an IC50 of 0.34 μM.

  • Phenol-based VL285 analog
  • E3 ubiquitin ligase VHL ligand
  • Used in the synthesis of HaloPROTAC3
  • Degrades HaloTag7 fusion protein (IC50=0.34 μM)
  • Appearance: White to off-white solid
  • Purity: 98.56%
  • Molecular weight: 548.65
  • Molecular formula: C29H32N4O5S

Catalog No. 50-003-74421 Supplier Medchemexpress LLC Supplier No. 25MGHY151227
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VL285 Phenol is a phenol-based VL285 analog and an E3 ubiquitin ligase VHL ligand. It can be used in the synthesis of HaloPROTAC3, where it aids in degrading HaloTag7 fusion protein with an IC50 of 0.34 μM.

  • Phenol-based VL285 analog
  • E3 ubiquitin ligase VHL ligand
  • Used in the synthesis of HaloPROTAC3
  • Degrades HaloTag7 fusion protein (IC50=0.34 μM)
  • Appearance: White to off-white solid
  • Purity: 98.56%
  • Molecular weight: 548.65
  • Molecular formula: C29H32N4O5S

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.