Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC VL285 phenol | 1448188-69-9 | MFCD31746640 | 98.6% | 548.65 g/mol | C29H32N4O5S | 10 MG
SDP

Catalog No. 5000287550
Encompass_Preferred

VL285 Phenol is a phenol-based analog of a VHL E3 ligase ligand, provided as a high-purity research chemical for use as a PROTAC building block and for VHL recruitment studies. It has reported activity in degrading HaloTag7 fusion proteins (IC50 = 0.34 μM).

  • Phenol-based VHL ligand analog.
  • Suitable for PROTAC synthesis and VHL studies.
  • High purity (98.56%).
  • Molecular weight 548.65 g/mol; formula C29H32N4O5S.
  • Soluble in DMSO at 100 mg/mL.
  • Powder storage: -20°C (up to 3 years), 4°C (up to 2 years).

Catalog No. 50-002-87550 Supplier Medchemexpress LLC Supplier No. HY15122710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VL285 Phenol is a phenol-based analog of a VHL E3 ligase ligand, provided as a high-purity research chemical for use as a PROTAC building block and for VHL recruitment studies. It has reported activity in degrading HaloTag7 fusion proteins (IC50 = 0.34 μM).

  • Phenol-based VHL ligand analog.
  • Suitable for PROTAC synthesis and VHL studies.
  • High purity (98.56%).
  • Molecular weight 548.65 g/mol; formula C29H32N4O5S.
  • Soluble in DMSO at 100 mg/mL.
  • Powder storage: -20°C (up to 3 years), 4°C (up to 2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.