Learn More
Athens Research And Technology Inc VLDL LIPOPROTEINS CELL CULTURE
Supplier:  Athens Research And Technology Inc 1216221204TC1MG
Encompass_Preferred
Lipoproteins, Very Low Density, HP, Cell Culture: 1 MG; Liquid in 150 mM NaCl, pH 7.4, and 0.1% EDTA; MW 10-80,000,000; Endotoxin Level: <=10 EU/mg. Native; Purified from fresh, non-frozen human plasma in the Athens Research & Tech lab.
Catalog No. 50-489-874