Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7436MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: VLDLR/1337. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: HeLa or U-251 cells. Pancreas, Cerebellum, Heart or Skeletal Muscle. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human VLDLR fragment from c-terminal. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 143-161kDa.Chromosome: 9p24. Gene Symbol: VLDLR. Gene ID: 7436. Ensemble: ENSG00000147852. Uniprot: NP_001309154.
Catalog No. 50-188-5276