Learn More
Sino Biological VLDLR / VLDL Receptor Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 11075RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human VLDLR (rh VLDLR; Catalog#11075-H08H; NP_003374.3; Met 1-Ser 797). VLDLR specific IgG was purified by human VLDLR affinity chromatography.
Catalog No. 50-224-8176