Learn More
S2 Media vLogiX ™ (VSTM) Kits, 50/box
Catalog No. 502995960
Encompass_Preferred
vLogiX ™ (VTSM) is a non-selective and non-differential culture medium intended for the collection, conveyance, and storage of clinical specimens containing viral agents. vLogiX ™ (VTSM) is intended for use in standard laboratory procedures for culture and diagnostic assays. Each kit contains 1 tube of 3mL VTSM with borosilicate beads and 1 nasopharyngeal swab within a reclosable bag. Each box contains 50 kits.• Product meets ASM, CDC, and CLSI reference methods and performance criteria.• vLogiX™ should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5960 Supplier S2 Media Supplier No. 5312
Add to Cart
Add to Cart
vLogiX ™ (VTSM) is a non-selective and non-differential culture medium intended for the collection, conveyance, and storage of clinical specimens containing viral agents. vLogiX ™ (VTSM) is intended for use in standard laboratory procedures for culture and diagnostic assays. Each kit contains 1 tube of 3mL VTSM with borosilicate beads and 1 nasopharyngeal swab within a reclosable bag. Each box contains 50 kits.• Product meets ASM, CDC, and CLSI reference methods and performance criteria.• vLogiX™ should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.