Learn More
BioVendor, LLC VISININ-LIKE PROTEIN 1 HUMAN SHEEP POLYCLONAL ANTIBODY 0.1MG
SDP
Supplier:  BioVendor, LLC RD184119100
Encompass
Visinin-like Protein 1 (VLP-1), Type Polyclonal AntibodyApplicationsWestern blotting, ELISASource of AntigenE. coliHostsSheepPreparationThe antibody was raised in sheep by immunization with the recombinant Human Visinin-like protein 1.Amino Acid SequenceRecombinant Human Visinin-like protein 1, total 201 AA. MW: 23.4 kDa (calculated). N-terminal His-tag, 10 extra AA (highlighted). The AA sequence is identical to UniProtKB/Swiss-Prot entry P62760.MKHHHHHHASMGKQNSKLAPEVMEDLVKSTEFNEHELKQWYKGFLKDCPSGRLNLEEFQQLYVKFFPYGDASKFAQHAFRTFDKNGDGTIDFREFICALSITSRGSFEQKLNWAFNMYDLDGDGKITRVEMLEIIEAIYKMVGTVIMMKMNEDGLTPEQRVDKIFSKMDKNKDDQITLDEFKEAAKSDPSIVLLLQCDIQKSpecies ReactivityHuman. Not yet tested in other species.Purification MethodImmunoaffinity chromatography on a column with immobilized recombinant Human Visinin-like protein 1.Antibody Content0.1 mg (determined by BCA method, BSA was used as a standard)
Catalog No. 50-412-7