Learn More
ACROBiosystems Fluorescent VLP isotype control (VLP-NF2P4) is expressed from human 293 cells (HEK293).
Catalog No. 502253937
Encompass_Preferred
Virus-like particles (VLPs) are highly structured protein particles composed of single or multiple structural proteins of the virus. Its morphological structure is similar to that of natural virus particles, does not contain virus accounting, and has strong immunogenicity and biological activity. The empty shell structure of most VLP is icosahedral or helical structure, which is composed of several specific structural proteins. At present, it is widely used in vaccine production and antigen presentation.
This item is not returnable. View return policy
Catalog No. 50-225-3937 Supplier ACROBiosystems Supplier No. VLPNF2P4150UG
Virus-like particles (VLPs) are highly structured protein particles composed of single or multiple structural proteins of the virus. Its morphological structure is similar to that of natural virus particles, does not contain virus accounting, and has strong immunogenicity and biological activity. The empty shell structure of most VLP is icosahedral or helical structure, which is composed of several specific structural proteins. At present, it is widely used in vaccine production and antigen presentation.